Learn More
Fisherbrand™ SureOne™ Reload Empty Racks
For use with Fisherbrand SureOne universal pipet tips
$96.00 - $112.29
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
02-707-443
|
N/A | 1250 μL |
Pack of 10 Each for $99.71
Case of 5 Pack for $498.55
|
|
|||||
|
02-707-444
|
N/A | 200 to 300 μL |
Pack of 10 Each for $112.29
Case of 5 Pack for $561.45
|
|
|||||
|
02-707-445
|
N/A | 10 to 20 μL |
Pack of 10 Each for $96.00
Case of 5 Pack for $480.00
|
|
|||||
Description
- Inserts (not included) sit flush with rack, allowing easy multichannel pipetting
- Notched clasp designed to prop lid open for autoclave venting
- Sure stack lid rim prevents racks from sliding around when stacked and carted around a lab
- Safe lock latch allows for one-handed opening and closing ability—won't pop open when dropped
- Sure snap design securely holds insert in place, preventing it from being lifted out while pipetting
- Flip-lid can be either a hinged or lift-off lid; hinged format does not prevent use of a multichannel pipet
Order Info
Inserts not included; must be ordered separately.
Specifications
| Pipette Tips | |
| Empty Rack |
As part of our commitment to continuous product improvement, your delivery may contain products with some minor color variation, but their form, fit and function will not be affected. Any questions contact your sales representative.