missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ SureOne™ Micropoint Pipette Tips, Universal Fit, Non-Filtered
GREENER_CHOICE

Catalog No. p-161433
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
100 to 1250 μL
5 to 300 μL
Sterility:
Non-sterile
Sterile
Tip Style:
Beveled
Extended Length
Micro Point
Format:
Bulk
Hinged Rack
Refill
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Tip Style Format
02707400 100 to 1250 μL Non-sterile Micro Point Bulk
02707407 100 to 1250 μL Sterile Micro Point Hinged Rack
02707408 100 to 1250 μL Non-sterile Micro Point Refill
02707409 5 to 300 μL Non-sterile Beveled Hinged Rack
02707410 5 to 300 μL Sterile Beveled Hinged Rack
02707447 5 to 300 μL Non-sterile Micro Point Bulk
02707411 5 to 300 μL Non-sterile Beveled Refill
02707436 0.1 to 10 μL Non-sterile Micro Point Hinged Rack
02707438 0.1 to 10 μL Non-sterile Micro Point Refill
02707440 0.1 to 10 μL Non-sterile Extended Length Hinged Rack
02707441 0.1 to 10 μL Sterile Extended Length Hinged Rack
02707454 0.1 to 10 μL Non-sterile Micro Point Bulk
02707437 0.1 to 10 μL Sterile Micro Point Hinged Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 02707400

New, improved Fisherbrand SureOne Reload system offers a full line of micropoint universal fit pipet tips

Reload system provides a complete line of micropoint universal-fit pipet tips.

  • All racked sterile tips are shrink wrapped and certified free of RNase, DNase, ATP, pyrogen, bioburden, PCR inhibitors and endotoxin and DNA All nonsterile racked and bulk nonfiltered tips are certified free of RNase, DNase, and DNA
SureOne Rack Features
  • Flush Fit Inserts sit flush with rack, allowing easy multichannel pipetting
  • Recycle Logo molded into all rack components
  • Notched clasp props lid open for autoclave venting
  • Sure Stack lid rim prevents racks from sliding around when stacked and carted around a lab
  • Safe-Lock latch allows for one-handed open and closing ability and won't pop open when dropped
  • Sure-snap design securely holds insert in place, preventing it from being lifted out while pipetting
  • Reversible flip-lid can be either a hinged or lift-off lid; hinged format does not prevent use of a multichannel pipet
SureOne Insert Features
  • Inserts are easy to place onto and remove from rack; clip onto rack for a secure fit
  • Lot traceable—all inserts contain the Fisherbrand part number, tip volume, lot number
  • Indented insert holes inhibit tip movement, stabilizing them for multichannel work
  • Contact Clip prevents unnecessary contact with the tips while clipping in inserts
  • Multichannel design; all inserts are 96-well format Robust non-pliable design provides secure seals on every tip
SureOne Reload Packaging Features
  • Product information displayed on box
  • Dust Flap prevents debris from contaminating the product
  • Finger Knockouts allow easy access to slide out product
  • Canyon Cut Package allows for easy tip insert removal
  • Reload packaging design saves 58% bench space vs. racked packaging; reduces plastic waste by 52%

Order Info

Packaging options include bulk, rack, rack sterile and reload.

Specifications

Volume (Metric) 100 to 1250 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Length (English) 3.5 in.
Length (Metric) 89 mm
Clean Claims DNase, RNase, and DNA Free
Format Bulk
Tip Style Micro Point
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line SureOne
Material Polypropylene
Graduations 100, 200, 500, 1000μL
Show More Show Less

As part of our commitment to continuous product improvement, your delivery may contain products with some minor color variation, but their form, fit and function will not be affected. Any questions contact your sales representative.