missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ SureOne™ Micropoint Nano Pipette Tips, Universal Fit

For use with micro volume pipetters
Description
For use with micro volume pipetters
- Natural color
- Precision molded for accuracy in low volume pipetting
- Certified by independent laboratory – Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and pyrogen-free
Chemical Identifiers
| Natural | |
| Micro Point | |
| SureOne |
Specifications
| 0.1 to 10 μL | |
| Natural | |
| 3.17cm | |
| Hinged Rack | |
| Universal Fit |
| Non-sterile | |
| 1.25 in. | |
| DNase, RNase, DNA, and Pyrogen Free | |
| Micro Point | |
| SureOne |