Learn More
Fisherbrand™ SureOne™ Micropoint Nano Pipette Tips, Universal Fit
GREENER_CHOICE
For use with micro volume pipetters
 View more versions of this product
Catalog No. 211972E
Only null left
Description
Description
For use with micro volume pipetters
  • Natural color
  • Precision molded for accuracy in low volume pipetting
  • Certified by independent laboratory – Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and pyrogen-free
Specifications
Chemical Identifiers
Natural
Micro Point
SureOne
Specifications
0.5 to 10 μL
Natural
4.5cm
Hinged Rack
Universal Fit
Non-sterile
1.81 in.
DNase, RNase, DNA, and Pyrogen Free
Micro Point
SureOne
Product Suggestions
Product Suggestions