missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ SureGrip™ Pipet Tip Reload System: Empty Racks

Catalog No. 05408195
Change view
Click to view available options
For Use With (Equipment):
10μL Long Reach, 200μL Tips
For 10μL tips
For 1250μL tips
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
05408195 For 1250μL tips
05408185 For 10μL tips
05408193 10μL Long Reach, 200μL Tips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05408195
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Empty racks for use with Fisherbrand SureGrip Pipet Tip Reload System.

  • Certified DNase, RNase, Human DNA and PCR inhibitor free
  • Color: Natural

Specifications

Certifications/Compliance Certified DNase, RNase, human DNA and PCR inhibitor free
Clean Claims DNase, RNase, DNA, and PCR inhibitor Free
Color Natural
For Use With (Equipment) For 1250μL tips
Length (Metric) 10.3 cm
Length (English) 4.05 in.
Product Type Empty Cell Rack