Learn More
Fisherbrand™ High-Speed Mini-Centrifuge, 4 x 8 Strip 0.2 mL Tube (with knob), Black
GREENER_CHOICE
Catalog No. 12006908
Change view
Click to view available options
For Use With (Equipment):
Gusto™ High Speed Mini Centrifuge
Catalog No. For Use With (Equipment)
12006908 Gusto™ High Speed Mini Centrifuge
1 options
Catalog No. 12006908 Supplier Fisherbrand™ Supplier No. HS100504
Strip Tube Rotor for High-Speed Mini-Centrifuge.
  • Strip Tube Rotor designed for use with Fisherbrand™ High-Speed Mini-Centrifuge
Specifications
Material Polycarbonate
Type Strip Tube Rotor
Capacity 32 Tubes
Max. RCF None
Max. Speed None
For Use With (Equipment) Gusto™ High Speed Mini Centrifuge
Includes Knob
Biocontainment No