missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Split Cowhide Leather Palm Gloves

Catalog No. p-2363585
Change view
Click to view available options
Glove Cuff Style:
Gauntlet
Safety
Size:
Large
Catalog No. Glove Cuff Style Size
19053924 Gauntlet Large
19013466 Safety Large
2 options

Catalog No. 19053924

Supplier: Fisherbrand™ 66GL

Only null left

Gloves feature a traditional fabric striped back with a split cowhide leather palm.

  • Traditional striped fabric back and cuff
  • Split cowhide leather palm
  • Leather pull patch
  • Shirred elastic back
  • Wing thumb
  • Available with a gauntlet or safety cuff

Specifications

Color Gray, Blue
Grade Standard
Hand Specific
Material Cowhide Leather
Product Type General Purpose Gloves
Size Large
Style Gunn
Glove Cuff Style Gauntlet
Hem Color Brown
Liner Material Cotton
Length (English) 12.3 in.
Length (Metric) 31.3 cm
Show More Show Less