Learn More
Fisherbrand™ Staining Control Slides
Description
- Red box on the slide identifies the control tissue
- Frosted area provides a positive marking spot
- Superfrost™ Plus treatment secures tissue to the slide throughout the staining process
- Cat. No. 22-042-910 has an extended frosted area, allowing room for the addition of patient information and/or a bar code label
Specifications
Specifications
| Type | Staining Control Slide |
| Description | Standard Frosted Area |
| For Use With (Application) | For automatic stainers |