Learn More
Fisherbrand™ Staining Control Slides
Catalog No. 22042910
Change view
Click to view available options
For Use With (Application):
For automatic stainers
For manual staining methods
Description:
Extended Frosted Area
Standard Frosted Area
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Description
22042910 For automatic stainers Extended Frosted Area
12518105 For automatic stainers Standard Frosted Area
22035819 For manual staining methods Standard Frosted Area
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22042910
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Allow staining of control and patient samples on the same slide with the same protocol
  • Red box on the slide identifies the control tissue
  • Frosted area provides a positive marking spot
  • Superfrost™ Plus treatment secures tissue to the slide throughout the staining process
  • Cat. No. 22-042-910 has an extended frosted area, allowing room for the addition of patient information and/or a bar code label
Specifications
Type Staining Control Slide
Description Extended Frosted Area
For Use With (Application) For automatic stainers