Learn More
Fisherbrand™ Square Labels
GREENER_CHOICE
Fisher branded Cryogenic Storage Labels use a special adhesive that is designed to withstand extreme cold temperatures as low as -196º C (-311º F). Our cryogenic storage labels can be printed or written on and will not bleed, smudge or smear. Engineered for you to use in your laser printer.
 View more versions of this product
Catalog No. 11010258
Only null left
Description
Description
  • Labels can be printed or written on and will not bleed, smudge or smear
  • Engineered for use in laser printers
Specifications
Chemical Identifiers
Non-sterile
Specifications
Orange
Square Label
1000/Roll
1.11 x 2.54 cm
Roll
Non-sterile
0.4375 x 1 in.
Product Suggestions
Product Suggestions