Learn More
Fisherbrand™ Square Labels
Fisher branded Cryogenic Storage Labels use a special adhesive that is designed to withstand extreme cold temperatures as low as -196º C (-311º F). Our cryogenic storage labels can be printed or written on and will not bleed, smudge or smear. Engineered for you to use in your laser printer.
Description
- Labels can be printed or written on and will not bleed, smudge or smear
- Engineered for use in laser printers
Chemical Identifiers
| Non-sterile |
Specifications
| Blue | |
| Square Label | |
| 1000/Roll | |
| 1.11 x 2.54 cm |
| Roll | |
| Non-sterile | |
| 0.4375 x 1 in. |