Learn More
Fisherbrand™ Square Labels
GREENER_CHOICE
Catalog No. 11010255
Change view
Click to view available options
Color:
Assorted
Blue
Green
Orange
Red
White
Yellow
Product Type:
Square Label
Quantity:
1000/Roll
156/Pk.
2000/Roll
5000/Roll
52/Sheet
85 Labels/Sheet
Size (Metric):
0.64 x 3.17 cm
0.64 x 3.81 cm
0.64 x 3.81cm
1.11 x 2.54 cm
1.27 x 3.17 cm
1.59 x 3.49 cm
1.91 x 2.54 cm
1.91 x 4.29 cm
26 product options available for selection
Product selection table with 26 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type Quantity Size (Metric)
11010255 Orange Square Label 1000/Roll 1.27 x 3.17 cm
11010209 White Square Label 52/Sheet 1.91 x 4.29 cm
11010210 Green Square Label 156/Pk. 0.64 x 3.81 cm
11010211 Orange Square Label 156/Pk. 0.64 x 3.81 cm
11010212 Red Square Label 156/Pk. 0.64 x 3.81cm
11010213 Yellow Square Label 156/Pk. 0.64 x 3.81cm
11010215 Assorted Square Label 156/Pk. 0.64 x 3.81 cm
11010214 White Square Label 156/Pk. 0.64 x 3.81cm
11010222 Orange Square Label 85 Labels/Sheet 0.64 x 3.17 cm
11010220 Blue Square Label 85 Labels/Sheet 0.64 x 3.17 cm
11010221 Green Square Label 85 Labels/Sheet 0.64 x 3.17 cm
11010223 Red Square Label 85 Labels/Sheet 0.64 x 3.17 cm
11010224 Yellow Square Label 85 Labels/Sheet 0.64 x 3.17 cm
11010242 White Square Label 1000/Roll 0.64 x 3.81 cm
11010245 White Square Label 2000/Roll 0.64 x 3.81 cm
11010243 White Square Label 1000/Roll 1.59 x 3.49 cm
11010256 Blue Square Label 1000/Roll 1.11 x 2.54 cm
11010257 Green Square Label 1000/Roll 1.11 x 2.54 cm
11010258 Orange Square Label 1000/Roll 1.11 x 2.54 cm
11010259 Red Square Label 1000/Roll 1.11 x 2.54 cm
11010260 White Square Label 1000/Roll 1.11 x 2.54 cm
11010261 Yellow Square Label 1000/Roll 1.11 x 2.54 cm
11010265 White Square Label 5000/Roll 1.91 x 2.54 cm
11010262 White Square Label 2000/Roll 1.11 x 2.54 cm
11010263 White Square Label 5000/Roll 1.11 x 2.54 cm
11010266 White Square Label 2000/Roll 1.27 x 3.17 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 26 options available.
26 options
Catalog No. 11010255
Only null left
Fisher branded Cryogenic Storage Labels use a special adhesive that is designed to withstand extreme cold temperatures as low as -196º C (-311º F). Our cryogenic storage labels can be printed or written on and will not bleed, smudge or smear. Engineered for you to use in your laser printer.
  • Labels can be printed or written on and will not bleed, smudge or smear
  • Engineered for use in laser printers
Specifications
Color Orange
Format Roll
Product Type Square Label
Sterility Non-sterile
Quantity 1000/Roll
Size (English) 0.5 x 1.25 in.
Size (Metric) 1.27 x 3.17 cm