Learn More
Fisherbrand™ Spherical Dewar Flask Fiberglass Caddie
Catalog No. 101965
Encompass
Change view
Click to view available options
Material:
Fiberglass
Catalog No. Material
101965 Fiberglass
1 options
Catalog No. 101965 Supplier Fisherbrand™ Supplier No. POS 30380
Safe method for carrying and decanting liquids on premises
  • Accommodates flask with standard aluminum base attached
  • Gaskets center the flask and hold it firmly in place
  • Reusable
  • Sturdy, lightweight construction
  • Side handle and cap
Specifications
Autoclavable Not Autoclavable
Color Blue
Material Fiberglass
Product Type Dewar Caddie Rack