missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Potassium Iodide Starch Test

Catalog No. 14860
Change view
Click to view available options
Quantity:
24 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14860 24 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14860 Supplier Fisherbrand™ Supplier No. 16024V100

Test for the presence of free iodine, chlorine and peroxides in solution

Quickly and easily test for the presence of free iodine, chlorine and peroxides in solution

  • The Potassium Iodide Starch test paper detects available chlorine, iodine and peroxides in the solution
  • Chlorine and Peroxides react immediately, while Iodine requires approximately 60 seconds
  • Initial reactions show a slight blue color, while higher concentrations turn the strip from dark blue to purple
  • The upper limits for chlorine and peroxides are between 400 to 450 ppm, and the upper limits for Iodine are at 50 to 100 ppm

Order Info

100 Strips per vial

Specifications

Color Indicator Blue to Deep Purple
Format Strip
For Use With (Application) Use this strip to test for the presence of free Iodine, Chlorine, and Peroxides in a solution
Includes 100 Test Strips/Vial
Quantity 24 Pack
Range The upper limits for chlorine and peroxides are between 400 to 450 ppm, and the upper limits for Iodine are at 50 to 100 ppm.
Test Time Chlorine and Peroxides react immediately, while Iodine requires approximately 60 seconds
Type Potassium Iodide Starch Test