Learn More
Fisherbrand™ Spatula with Stainless-Steel Blade
Fisherbrand Spatula with Stainless-Steel Blade are non-stick, corrosion-free with a bright finish.
Description
- Fisherbrand™ Spatula with Stainless-Steel Blade are non-stick, corrosion-free with a bright finish
- Multipurpose - Useful in educational, research or healthcare applications
- Multiple sizes allow for varied laboratory applications
- Made from ASTM stainless steel
- Non-sticking, corrosion-free standard blade
- Riveted, smooth hardwood handle designed and engineered to fit comfortably in your hand
- Electronically passivated for corrosion resistance
- Autoclavable
- Latex-free
Chemical Identifiers
| Spatula | |
| Stainless Steel | |
| Riveted |
| Bright | |
| Hardwood |
Specifications
| Spatula | |
| 10 in. | |
| Stainless Steel | |
| Riveted |
| Bright | |
| 25.4 cm | |
| Hardwood |