missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Solid Rubber Stoppers

Available in a variety of sizes ranging from 000 through 16

Manufacturer:  Fisherbrand™ 6M290

Catalog No. 14-130J




Description

Description

Available in a variety of sizes ranging from 000 through 16

  • Solid rubber construction

The approximate per-pack quantity shown is based on the average number of stoppers in 1 lb. (454g).

Specifications

Specifications

2.6cm
General purpose use
Solid Taper Stopper
6
Natural Rubber
3.2cm
2.5cm
6
Black
Documents