missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Small Square Biohazard Labels
GREENER_CHOICE

Catalog No. 18999930
Change view
Click to view available options
Color:
Black, Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18999930 Black, Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18999930 Supplier Fisherbrand™ Supplier No. LBH 18

Bright red fluorescent labels display the universal biohazard symbol for easy identification.

  • Strong adhesive ensures that labels remain in place
  • Labels with black 1 in. core

Compliance

Meet OSHA requirements.

Specifications

Certifications/Compliance OSHA
Color Black, Red
Format Label
Material Paper
Product Type Square Biohazard Label
For Use With (Application) Biohazard
Quantity 1000/Roll
Shape Square
Size (English) 0.93 x 0.93 in.
Size (Metric) 2.36 x 2.36 cm