missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ SlideRite™ Cytology Fixative

Catalog No. 0435588
Change view
Click to view available options
Quantity:
2 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
0435588 2 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0435588 Supplier Fisherbrand™ Supplier No. 1801

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A water soluble fixative for use in exfoliative cytology

  • Forms a clear protective coating over specimens and fixates cells immediately
  • Protects samples during transportation and while in storage
  • Dries in as little as five minutes

Specifications

For Use With (Equipment) Microscope
Grade Chemical Solution
Product Type Fixative Spray
Quantity 2 oz.
Solution Type Fixative
For Use With (Application) Protective coating over all types of cytological smears
Form Liquid