Learn More
Fisherbrand™ Siteliner Safety Glasses, Blue frame
Unilens design offers wide field of vision and exceptional clarity
$50.24 - $50.24
Specifications
Lens Coating Hardcoat
Certifications/Compliance Meets ANSI Z87.1-2003 standards.
Lens Tint Clear
Lens Material Polycarbonate
View More Specs
Products 1
Catalog Number Mfr. No. Frame Color Price Quantity  
Catalog Number Mfr. No. Frame Color Price Quantity  
19-130-2095
missing translation for 'viewDocuments'
Fisherbrand™
191302095
Blue
Pack of 12 Each for $50.24
Case of 10 Pack for $487.00
Only null left
Description
Description
Unilens design offers wide field of vision and exceptional clarity
  • Lightweight, impact-resistant nylon frames
  • One-piece, clear hardcoat polycarbonate lens for maximum protection
  • Scratch resistant; absorb >99.9% of harmful UV light
  • Molded-in sideshields and browguards for added protection against UV, dust and flying particles
  • Adjustable temples and lens inclination
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Specifications
Hardcoat
Meets ANSI Z87.1-2003 standards.
Clear
Polycarbonate