Learn More
Fisherbrand™ Serialized Class A Cylinders with Certificate of Calibration
Calibrated To Deliver.
 Afficher plus de versions de ce produit
Numéro de catalogue. 08-557EC
Il en reste null
Découvrez plus d'offres spéciales
Comprend: Individual fused-in serial numbers and, in addition to traceability certification, are supplied with Certificates of Calibration that list apparent volumes and calibration correction factors
Description
Description
Calibrated To Deliver.
  • Traceability
  • White scale
Spécifications
Spécifications
Round
Graduated Cylinder
To Deliver
Hexagonal
330 mm
250 mL
2 mL
±1 mL
12.9 in.
White
A
Borosilicate Glass
Individual Fused-In Serial Numbers, Traceable NIST Certificate
2mL
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Accessoires