missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Self-Closing Artery Forceps

Catalog No. p-128845
Change view
Click to view available options
Tip Style:
Straight, serrated, blunt point
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Style
138124 Straight, serrated, blunt point
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 138124

Fisherbrand Self-Closing Artery Forceps are cross-actioned forceps primarily used to control blood flow. ASTM™ standard (F899-84) stainless steel.

  • Fisherbrand™ Self-Closing Artery Forceps are cross-actioned forceps primarily used to control blood flow
  • Our forceps are designed to exceed perfromance expectations -This is the Fisherbrand Advantage.
  • Automatic clamping - squeeze patterned handle to open
  • Designed for comfort, stability and ease of use
  • Multipurpose - Useful in educational, research or healthcare setting
  • Hand polished to mirror finish
  • Dieffenbach Serrefines
  • Made from surgical stainless steel
  • Electronically passivated for corrosion resistance
  • Can be sterilized by Eto/gamma radiation
  • Rockwell hardness (HRC): 40-45
  • Autoclavable up to 135°C
  • Latex-free

Certifications

Manufactured under ISO 9002, CGMP guidelines.

Specifications

Product Type Artery Forcep
Autoclavable Autoclavable
Certifications/Compliance ISO 9002, cGMP
Material Stainless Steel
Tip Style Straight, serrated, blunt point
Length (English) 2.25 in.
Length (Metric) 5.7 cm