Learn More
Fisherbrand™ Screw Top Polypropylene Histology Containers with Lids
Convenient leak-resistant histology containers in assorted sizes.
Supplier:  Fisherbrand™ ES60240F
 View more versions of this product
Catalog No. 22026312
Only null left
Description
Description
Maintain the integrity of your sample during transport and storage. 
  • High-quality, clarified polypropylene containers
  • Secure, leak-resistant screw cap closures
  • Available in various sizes to hold from small to large specimens
  • Larger volume sizes feature a wide mouth opening
Note: Containers are autoclavable; not caps or cap liners.
Specifications
Chemical Identifiers
Polypropylene (Clarified)
HDPE
Leak-resistant Screw Cap
Specifications
Polypropylene (Clarified)
HDPE
24/Cs.
8 oz.
Leak-resistant Screw Cap
Product Suggestions
Product Suggestions