Learn More
Fisherbrand™ Safety Blue Thermohydrometer, Fahrenheit
Lead and mercury free thermohydrometers.
Description
Hydrometer with enclosed thermometer to indicate temperature of the liquid under test.
Chemical Identifiers
| NIST | |
| Lead and Mercury Free |
Specifications
| Thermo Hydrometer | |
| 0.1 | |
| 385 mm | |
| Lead and Mercury Free |
| 6H | |
| NIST | |
| Soda Lime Glass | |
| 69 to 81 |