Learn More
Fisherbrand™ Safety Blue Thermohydrometer, Fahrenheit
Lead and mercury free thermohydrometers.
 View more versions of this product
Catalog No. 11555226
Only null left
Description
Description
Hydrometer with enclosed thermometer to indicate temperature of the liquid under test.
Specifications
Chemical Identifiers
NIST
Lead and Mercury Free
Specifications
Thermo Hydrometer
0.1
385 mm
Lead and Mercury Free
6H
NIST
Soda Lime Glass
49 to 61
Product Suggestions
Product Suggestions