Learn More
Fisherbrand™ Safety Blue Thermohydrometer, Fahrenheit
Lead and mercury free thermohydrometers.
 View more versions of this product
Catalog No. 11555223
Only null left
Description
Description
Hydrometer with enclosed thermometer to indicate temperature of the liquid under test.
Specifications
Chemical Identifiers
NIST
Lead and Mercury Free
Specifications
Thermo Hydrometer
0.1
385 mm
Lead and Mercury Free
6H
NIST
Soda Lime Glass
29 to 41
Product Suggestions
Product Suggestions