Learn More
Fisherbrand™ Safety Blue Thermohydrometer, Celsius
Lead and mercury free
Description
- Hydrometer with enclosed thermometer to indicate temperature of the liquid under test.
Chemical Identifiers
| Thermo Hydrometer | |
| NIST | |
| Lead and Mercury Free | |
| For determining the percentage by weight of sucrose |
Specifications
| Thermo Hydrometer | |
| 79 to 91° | |
| For determining the percentage by weight of sucrose |
| NIST | |
| Lead and Mercury Free |