Learn More
Fisherbrand™ SafeSnap™ Disposable Ampule Openers
Polyethylene ampule collars protect fingers while breaking open the scored neck of glass ampules.
Specifications
| Disposable | Single-use |
|---|---|
| Product Type | Ampule Opener |
| Material | Polyethylene |
Description
Polyethylene ampule collars protect fingers while breaking open the scored neck of glass ampules.
Polyethylene ampule collars protect fingers while breaking open the scored neck of glass ampules.
- To use: Slip the collar onto the ampule until collar rests on the widest part
- Grip the collar with thumb and forefinger while holding the upright ampule at bottom
- Break the ampule with a quick snap
- Dispose of the collar and ampule top together
Specifications
| Single-use | |
| Ampule Opener | |
| Polyethylene |