Learn More
Fisherbrand™ Round Covered Lead Flask Rings
Stabilize Erlenmeyer flasks immersed in water baths.
$80.62 - $114.90
Specifications
Autoclavable Not Autoclavable
Color Blue
Material Lead
Positions 1
Shape Round
View More Specs
Products 4
Catalog Number Diameter (Metric) Inner Weight (Metric) Price Quantity & Availability  
Catalog Number Diameter (Metric) Inner Weight (Metric) Price Quantity & Availability  
058696
View Documents Promotion Details
5.1 cm 453.5 g
Each for $95.93
Only null left
058695
View Documents Promotion Details
4.1 cm 226.7 g
Each for $80.62
Only null left
058697
View Documents Promotion Details
5.6 cm 680.3 g
Each for $106.52
Only null left
058698
View Documents Promotion Details
7 cm 907.1 g
Each for $114.90
Only null left
Description
Description
Stabilize Erlenmeyer flasks immersed in water baths.
  • Reduces floating, breakage and chemical spills
  • Bright blue coating
  • Chemical-resistant
  • Will not scratch flasks
  • Available in four diameter/weight combinations
Specifications
Specifications
Not Autoclavable
Lead
Round
Lead Ring
Blue
1
Non-sterile
None