Learn More
Fisherbrand™ Reusable Glass Tubes with Phenolic Screw Caps
Phenolic screw cap with white rubber liner
 View more versions of this product
Catalog No. 1495925B
Only null left
Description
Description
Phenolic screw cap with white rubber liner
  • High-quality borosilicate glass
  • Marking spot
  • Autoclavable
Order Info
Packed in shrink-wrapped, lint-free trays
Specifications
Chemical Identifiers
Phenolic
Suitable for tissue culture work and general bacteriological use
Rubber
Autoclavable
Reusable Glass Tube
Borosilicate Glass
Marking Spot
Non-sterile
Screw Cap
Specifications
Phenolic
100 mm
Suitable for tissue culture work and general bacteriological use
Rubber
Non-sterile
16 mm
Reusable Glass Tube
12 mL
Borosilicate Glass
Marking Spot
15-415
Autoclavable
Screw Cap
Round
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories