Learn More
Fisherbrand™ Reusable Glass Class A Volumetric Flasks with Snap Caps
Description
Includes
- 28150R polyethylene snap cap
Class A accuracy–without a serial number
- Calibrated “To Contain”
Compliance
- Designed from ASTM™ specification E288
Specifications
Specifications
| Calibration | Calibrated To Contain |
| Certifications/Compliance | Class A, ASTM E288 |
| Closure Material | Polyethylene |
| Capacity (Metric) | 50 mL |
| Material | Glass |
| No. per Case | 1 |
| Graduated | Yes |
| Closure Size | 16 |