missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Repeating Pipet Filler/Dispenser

Catalog No. 1368125
Change view
Click to view available options
Fits pipets up to 10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
1368125 Fits pipets up to 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1368125

Fits pipets up to 10mL.

  • Molded PVC body with rubber bellows
  • Depress plunger to expel air
  • Release plunger to draw up liquid; depress to release
  • Control cap adjusts plunger stroke for repeating manually preset volume up to 10mL

Specifications

Autoclavable Not Autoclavable
Color Yellow/Black
Material PVC, Rubber
Type Repeating Pipette Filler/Dispenser
For Use With (Application) Pipets up to 10mL
For Use With (Equipment) 10mL