missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Redi-Tip™ Pipet Tips: General Purpose
GREENER_CHOICE

Catalog No. p-4909390
Change view
Click to view available options
Volume (Metric):
1 to 200 μL
100 to 1000 μL
Tip Style:
Universal
Universal Fit
Color:
Blue
Natural
Yellow
Format:
Bulk
Hinged Rack
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Tip Style Color Format
02707502 1 to 200 μL Universal Fit Yellow Hinged Rack
02707511 100 to 1000 μL Universal Natural Hinged Rack
02707507 100 to 1000 μL Universal Blue Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02707502
Only null left

Premium virgin polypropylene resin and advanced molding methods produce a superior, high-quality pipet tip

  • Certified by independent laboratory
  • Nonsterile: RNase, DNase, DNA, and pyrogen free
  • Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and pyrogen free

  • For use with most 200 and 1000μL single and multichannel pipetters
  • Hinged racks can be stacked rack-on-rack
  • Hinged closure on racks allows top lid to be partially open for steam penetration during autoclaving

Packaging Code -- BP: Bulk Pack; RI: Rack Inserts; HR: Hinged Rack; HRS: Hinged Rack, Sterile; HR/T: Hinged Rack, Trace Metal-free; LR: Lift-off Rack; LRS: Lift-off Rack, Sterile; SR: Stack Rack; IWS: Individually Wrapped, Sterile.

Specifications

Volume (Metric) 1 to 200 μL
Sterility Sterile
Color Yellow
Autoclavable Autoclavable
Length (English) 2 in.
Length (Metric) 5cm
Clean Claims DNase, RNase, DNA, Bioburden, and PCR inhibitor Free
Format Hinged Rack
Tip Style Universal Fit
For Use With (Equipment) Universal Fit
Product Line Redi-Tip
Material Polypropylene
Show More Show Less