Learn More
Fisherbrand™ Quartz Ultramicro cells
Available with a variety of window materials for wide range of assays
Supplier:  Fisherbrand™ 105202QSZ85
 View more versions of this product
Catalog No. 14385928E
Only null left
Includes: Fitted PTFE covers or tapered PTFE stoppers
Description
Description
  • 8.5mm center height fits most Beckman™ Instruments and Spectronic™ spectrophotometers
  • 15.0mm cells fit most Hewlett-Packard™, Perkin-Elmer™, Shimadzu™, Unicam™ and GBC spectrophotometers Choice of correct center height is made such that optical aperture matches center position of light beam
Specifications
Chemical Identifiers
Quartz
Specifications
50 μL
Quartz
8.5 mm
Product Suggestions
Product Suggestions