missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Quartz Ultramicro cells

Catalog No. 14385928D
Change view
Click to view available options
Capacity (Metric):
50 μL
100 μL
Height (Metric):
8.5 mm
15 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Height (Metric)
14385928D 100 μL 15 mm
14385928E 50 μL 8.5 mm
14385928F 50 μL 15 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14385928D Supplier Fisherbrand™ Supplier No. 105201QS10Z15

Includes

Fitted PTFE covers or tapered PTFE stoppers

Available with a variety of window materials for wide range of assays

  • 8.5mm center height fits most Beckman™ Instruments and Spectronic™ spectrophotometers
  • 15.0mm cells fit most Hewlett-Packard™, Perkin-Elmer™, Shimadzu™, Unicam™ and GBC spectrophotometers Choice of correct center height is made such that optical aperture matches center position of light beam

Specifications

Capacity (Metric) 100 μL
Material Quartz
Height (Metric) 15 mm