missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ String Knit Gloves with PVC Dot Grip

Catalog No. 1138757
Change view
Click to view available options
Coating Material:
PVC Dots (1 Side)
PVC Dots (2 Sides)
Size:
Small
Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Coating Material Size
1138757 PVC Dots (1 Side) Small
19816783 PVC Dots (1 Side) Large
1138752 PVC Dots (2 Sides) Small
1138753 PVC Dots (2 Sides) Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 1138757 Supplier Fisherbrand™ Supplier No. 36110PD/S
Only null left

PVC dots enhance grip capabilities and abrasion resistance.

  • Cotton string knit gloves with PVC dots on either one side or both sides
  • Ideal for protecting hands from dirt and abrasion
  • Seamless construction is comfortable and reversible
  • Available in universal women's and men's sizes
  • Color-coded hem for quick size identification
  • Standard weight
  • Launderable

Specifications

Construction String Knit
Accelerator Free No
Applications Industrial
Coating Material PVC Dots (1 Side)
Color Natural, Black
Cut Resistance Rating ANSI A1
Disposable Reusable
Exterior Finish Grip Pattern
Hand Specific
Material Cotton
Powdered Powder Free
Product Type String Knit Gloves
Size Small
Sterility Non-sterile
Glove Cuff Style Continuous Knit
Grip Design Dots
Hem Color Yellow
Length (English) 8.9 in.
Length (Metric) 22.6 cm
Show More Show Less