missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Disposable Stir Bars

Catalog No. p-164481
Change view
Click to view available options
Length (Metric):
12.7 mm
25.4 mm
41.3 mm
50.8 mm
Diameter (Metric):
3.2 mm
8 mm
Length (English):
0.5 in.
1 in.
1.61 in.
2 in.
Diameter (English):
0.125 in.
0.31 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Length (English) Diameter (English)
1451383 50.8 mm 8 mm 2 in. 0.31 in.
1451393 12.7 mm 3.2 mm 0.5 in. 0.125 in.
1451395 41.3 mm 8 mm 1.61 in. 0.31 in.
1451394 25.4 mm 8 mm 1 in. 0.31 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 1451383
Only null left

The multifaceted magnetic stirring bar surface adds turbulence relative to similar size smooth cylindrical stirring bars.

  • Ideal for single use applications mixing radioactive reagents or when cross contamination might be a concern
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclavable

Specifications

Coating PTFE
Autoclavable Autoclavable
Length (Metric) 50.8 mm
Diameter (Metric) 8 mm
Material PTFE
Shape Cylinder
Type Stir Bar
Quantity 100
Certifications/Compliance ISO 9001:2008
Length (English) 2 in.
Color White
Diameter (English) 0.31 in.
Show More Show Less