missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Premium Grade Cowhide Leather Palm Work Gloves

Catalog No. 19053932
Change view
Click to view available options
Cuff Material:
Rubberized
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cuff Material
19053932 Rubberized
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19053932 Supplier Fisherbrand™ Supplier No. 827763/L

Gloves are constructed from premium grade cowhide leather with a cotton lined palm for additional comfort.

  • Premium grade cowhide leather construction
  • Cotton lined palm for comfort
  • Gunn cut pattern
  • Wing thumb

Specifications

Cuff Material Rubberized
Color Gray, Blue, Green
Grade Premium
Hand Specific
Material Cowhide Leather
Product Type General Purpose Gloves
Size Large
Style Gunn
Glove Cuff Color Blue with Green Stripes
Glove Cuff Style Safety
Hem Color Brown
Liner Material Cotton
Thumb Style Wing
Length (English) 10.5 in.
Length (Metric) 26.8 cm
Show More Show Less