missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Porcelain Spot Plates

For observing color reactions

$21.71 - $21.71

Specifications

Color White
Type Spot Plate
For Use With (Application) For observing color reactions
View More Specs

Products 1
Catalog Number Mfr. No. No. of Wells Dimensions (L x W) Price Quantity  
Catalog Number Mfr. No. No. of Wells Dimensions (L x W) Price Quantity  
FB964B
View Documents
N/A 12 4.4 x 3.6 in. (112 x 92 mm)
Each for $21.71
Case of 12 Each for $260.52
Only null left
 
Description

Description

  • Material: porcelain
  • Plate color: white
  • Available with 3 or 12 wells

Specifications

Specifications

White
Spot Plate
For observing color reactions
Videos
SDS
Documents

Documents