missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropylene Disposable Beakers

Catalog No. FB012917
Change view
Click to view available options
Capacity (Metric):
15 mL
30 mL
50 mL
100 mL
150 mL
250 mL
400 mL
600 mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
FB012917 100 mL
FB012918 15 mL
FB012919 30 mL
FB0129110 50 mL
FB0129111 150 mL
FB012915 250 mL
FB0129112 400 mL
FB012916 600 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. FB012917
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Highly resistant to strong acids, organic solvents and other common laboratory reagents

Rib construction permits stacking without binding

  • Special “dripless” lip
  • Base allows beaker to sit flat during use
  • Graduated in mL and oz.

Specifications

Capacity (Metric) 100 mL
Material Polypropylene
For Use With (Application) Highly Resistant to Strong Acids, Organic Solvents and Other Common Laboratory Reagents
Includes Spout
Increments 5 mL
Type Beaker
Autoclavable Autoclavable
Disposable Single-use
Graduated Yes