Learn More
Fisherbrand™ Polypropylene Disposable Beakers
Highly resistant to strong acids, organic solvents and other common laboratory reagents
 View more versions of this product
Catalog No. FB012916
Only null left
Description
Description
Rib construction permits stacking without binding
  • Special “dripless” lip
  • Base allows beaker to sit flat during use
  • Graduated in mL and oz.
Specifications
Chemical Identifiers
Polypropylene
Spout
Autoclavable
Yes
Highly Resistant to Strong Acids, Organic Solvents and Other Common Laboratory Reagents
Beaker
Single-use
Specifications
600 mL
Highly Resistant to Strong Acids, Organic Solvents and Other Common Laboratory Reagents
25 mL
Autoclavable
Yes
Polypropylene
Spout
Beaker
Single-use
Product Suggestions
Product Suggestions