missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Polypropylene Disposable Beakers
Description
Rib construction permits stacking without binding
- Special “dripless” lip
- Base allows beaker to sit flat during use
- Graduated in mL and oz.
Specifications
Specifications
| Capacity (Metric) | 400 mL |
| Material | Polypropylene |
| For Use With (Application) | Highly Resistant to Strong Acids, Organic Solvents and Other Common Laboratory Reagents |
| Includes | Spout |
| Increments | 25 mL |
| Type | Beaker |
| Autoclavable | Autoclavable |
| Disposable | Single-use |
| Graduated | Yes |