missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Polypropylene Disposable Beakers
Highly resistant to strong acids, organic solvents and other common laboratory reagents
Description
Rib construction permits stacking without binding
- Special “dripless” lip
- Base allows beaker to sit flat during use
- Graduated in mL and oz.
Chemical Identifiers
| Polypropylene | |
| Spout | |
| Autoclavable | |
| Yes |
| Highly Resistant to Strong Acids, Organic Solvents and Other Common Laboratory Reagents | |
| Beaker | |
| Single-use |
Specifications
| 150 mL | |
| Highly Resistant to Strong Acids, Organic Solvents and Other Common Laboratory Reagents | |
| 5 mL | |
| Autoclavable | |
| Yes |
| Polypropylene | |
| Spout | |
| Beaker | |
| Single-use |