Learn More
Fisherbrand™ Polyethylene Foam Surface Liners
Fisherbrand Polyethylene Foam Surface Liners protect against glass breakage in sinks and can be used as titration backgrounds, among hundreds of other uses around a laboratory.
 View more versions of this product
Catalog No. 1412749
Only null left
Description
Description
Fisherbrand Polyethylene Foam Surface Liners protect against glass breakage in sinks and can be used as titration backgrounds, among hundreds of other uses around a laboratory.
  • Cushion sensitive instruments and valuable glassware against vibration and breakage
  • Provide surface protection from chemicals, heat, dirt, and broken glass
  • Tape, staple, or tack in place
  • Easily cut to desired size
  • Easy to clean
  • No chlorofluorocarbon blowing agents used
  • White
Specifications
Chemical Identifiers
Surface Liner
Polyethylene Foam
White
Provides surface protection from chemicals, heat, dirt, and broken glass
Specifications
Surface Liner
Non-absorbent
15.2 m
457 mm
White
Provides surface protection from chemicals, heat, dirt, and broken glass
2 Case
Polyethylene Foam
50 ft.
18 in.
50 ft. x 18 in.
Single-use
0.12 in.
Roll
Product Suggestions
Product Suggestions