missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Autoclavable HD Biohazard Waste Bags

Catalog No. 01829D
Change view
Click to view available options
Color:
Clear
Clear with Red Print
Natural
Red with Black Print
Dimensions (L x W):
12 x 8 in. (30 x 20 cm)
19 x 14 in. (48 x 36 cm)
23 x 19 in. (58 x 48 cm)
24 x 12 in. (61 x 30 cm)
24 x 36 in. (61 x 91 cm)
25 x 35 in. (64 x 89 cm)
30 x 24 in. (76 x 61 cm)
30 x 33 in. (76 x 84 cm)
36 x 24 in. (91 x 61 cm)
48 x 38 in. (122 x 97 cm)
Thickness:
1.50 mil
4 mil
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Dimensions (L x W) Thickness
01829D Red with Black Print 25 x 35 in. (64 x 89 cm) 1.50 mil
018141 Natural 24 x 12 in. (61 x 30 cm) 1.50 mil
018143 Natural 24 x 36 in. (61 x 91 cm) 1.50 mil
01827A Clear with Red Print 12 x 8 in. (30 x 20 cm) 1.50 mil
01827D Clear with Red Print 36 x 24 in. (91 x 61 cm) 1.50 mil
01827B Clear with Red Print 24 x 12 in. (61 x 30 cm) 1.50 mil
01827C Clear with Red Print 30 x 24 in. (76 x 61 cm) 1.50 mil
01829A Red with Black Print 12 x 8 in. (30 x 20 cm) 1.50 mil
01829B Red with Black Print 19 x 14 in. (48 x 36 cm) 1.50 mil
01829C Red with Black Print 23 x 19 in. (58 x 48 cm) 1.50 mil
01829F Red with Black Print 30 x 33 in. (76 x 84 cm) 1.50 mil
01831A Red with Black Print 36 x 24 in. (91 x 61 cm) 4 mil
01832A Clear 36 x 24 in. (91 x 61 cm) 4 mil
018142 Clear 30 x 24 in. (76 x 61 cm) 1.50 mil
01829E Red with Black Print 48 x 38 in. (122 x 97 cm) 1.50 mil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 01829D Supplier Fisherbrand™ Supplier No. 1829D
Edge

missing translation for 'validMainframeMsg'

Autoclavable, polyethylene biohazard bags ensure safe handling and disposal of biohazardous waste.

Autoclavable, polyethylene biohazard bags to ensure safe handling and disposal of biohazardous waste.

  • All bags supplied with autoclavable elastic steam-venting closures except 1.5 mil-thick plain clear bags
  • Polyethylene
  • Temperature range 260°F
  • Available in 4 colors/styles

Specifications

Certifications/Compliance OSHA, 29 CFR1910.1030
Closure Type Elastic
Color Red with Black Print
Dimensions (L x W) 25 x 35 in. (64 x 89 cm)
Length (Metric) 63.5 cm
Width (Metric) 88.9 cm
Autoclavable Autoclavable
Includes Printed in English and Spanish
Width (English) 35 in.
Length (English) 25 in.
Thickness 1.50 mil
Material Polyethylene
Product Type Autoclavable Bags
Temperature (Metric) 127°C
Show More Show Less