missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Autoclavable HD Biohazard Waste Bags

Catalog No. 01829A
Change view
Click to view available options
Color:
Clear
Clear with Red Print
Natural
Red with Black Print
Dimensions (L x W):
12 x 8 in. (30 x 20 cm)
19 x 14 in. (48 x 36 cm)
23 x 19 in. (58 x 48 cm)
24 x 12 in. (61 x 30 cm)
24 x 36 in. (61 x 91 cm)
25 x 35 in. (64 x 89 cm)
30 x 24 in. (76 x 61 cm)
30 x 33 in. (76 x 84 cm)
36 x 24 in. (91 x 61 cm)
48 x 38 in. (122 x 97 cm)
Thickness:
1.50 mil
4 mil
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Dimensions (L x W) Thickness
01829A Red with Black Print 12 x 8 in. (30 x 20 cm) 1.50 mil
018141 Natural 24 x 12 in. (61 x 30 cm) 1.50 mil
018143 Natural 24 x 36 in. (61 x 91 cm) 1.50 mil
01827A Clear with Red Print 12 x 8 in. (30 x 20 cm) 1.50 mil
01827D Clear with Red Print 36 x 24 in. (91 x 61 cm) 1.50 mil
01827B Clear with Red Print 24 x 12 in. (61 x 30 cm) 1.50 mil
01827C Clear with Red Print 30 x 24 in. (76 x 61 cm) 1.50 mil
01829B Red with Black Print 19 x 14 in. (48 x 36 cm) 1.50 mil
01829C Red with Black Print 23 x 19 in. (58 x 48 cm) 1.50 mil
01829D Red with Black Print 25 x 35 in. (64 x 89 cm) 1.50 mil
01829F Red with Black Print 30 x 33 in. (76 x 84 cm) 1.50 mil
01831A Red with Black Print 36 x 24 in. (91 x 61 cm) 4 mil
01832A Clear 36 x 24 in. (91 x 61 cm) 4 mil
018142 Clear 30 x 24 in. (76 x 61 cm) 1.50 mil
01829E Red with Black Print 48 x 38 in. (122 x 97 cm) 1.50 mil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Numéro de catalogue. 01829A Fournisseur Fisherbrand™ Code fournisseur 1829A
Edge

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Autoclavable, polyethylene biohazard bags ensure safe handling and disposal of biohazardous waste.

Autoclavable, polyethylene biohazard bags to ensure safe handling and disposal of biohazardous waste.

  • All bags supplied with autoclavable elastic steam-venting closures except 1.5 mil-thick plain clear bags
  • Polyethylene
  • Temperature range 260°F
  • Available in 4 colors/styles

Spécifications

Certifications/Compliance OSHA, 29 CFR1910.1030
Closure Type Elastic
Color Red with Black Print
Dimensions (L x W) 12 x 8 in. (30 x 20 cm)
Length (Metric) 30 cm
Width (Metric) 20 cm
Autoclavable Autoclavable
Includes Printed in English and Spanish
Width (English) 8 in.
Length (English) 12 in.
Thickness 1.50 mil
Material Polyethylene
Product Type Autoclavable Bags
Temperature (Metric) 127°C
Afficher plus de résultats Afficher moins de résultats