missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Autoclavable HD Biohazard Waste Bags
Autoclavable, polyethylene biohazard bags ensure safe handling and disposal of biohazardous waste.
Supplier: Fisherbrand™ 1829E
Description
Autoclavable, polyethylene biohazard bags to ensure safe handling and disposal of biohazardous waste.
- All bags supplied with autoclavable elastic steam-venting closures except 1.5 mil-thick plain clear bags
- Polyethylene
- Temperature range 260°F
- Available in 4 colors/styles
Specifications
| OSHA, 29 CFR1910.1030 | |
| Red with Black Print | |
| 122 cm | |
| Autoclavable | |
| 38 in. | |
| 1.50 mil | |
| Autoclavable Bags |
| Elastic | |
| 48 x 38 in. (122 x 97 cm) | |
| 97 cm | |
| Printed in English and Spanish | |
| 48 in. | |
| Polyethylene | |
| 127°C |