missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Autoclavable HD Biohazard Waste Bags

Autoclavable, polyethylene biohazard bags ensure safe handling and disposal of biohazardous waste.

Supplier:  Fisherbrand™ 1832A

 View more versions of this product

Catalog No. 01-832A

Only null left
Explore available promotions

Description

Description

Autoclavable, polyethylene biohazard bags to ensure safe handling and disposal of biohazardous waste.

  • All bags supplied with autoclavable elastic steam-venting closures except 1.5 mil-thick plain clear bags
  • Polyethylene
  • Temperature range 260°F
  • Available in 4 colors/styles
Specifications

Specifications

ASTM D1709-85
Clear
91 cm
Autoclavable
36 in.
Polyethylene
127°C
Elastic
36 x 24 in. (91 x 61 cm)
61 cm
24 in.
4 mil
Autoclavable Bags
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions