missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Plastic pH Strips
Laboratory grade plastic pH strips in durable and easy to open flip-top vials
$33.94 - $37.57
Specifications
| For Use With (Application) | For quick and accurate matches |
|---|---|
| Quantity | 100 Strips/Vial, 6 Vials/Pk. |
| Storage Requirements | Room Temperature |
| Type | pH Strip |
| Catalog Number | Mfr. No. | pH Range | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | pH Range | Price | Quantity & Availability | |||||
|
13640520
|
Fisherbrand™
B60SHTRG000060VPS |
0 to 6 |
Each for $33.94
Pack of 6 Each for $203.64
|
|
|||||
|
13640516
|
Fisherbrand™
B60WIDRG000140VPS |
0 to 14 |
Pack of 100 Each for $37.57
Case of 6 Pack for $225.42
|
|
|||||
|
13640519
|
Fisherbrand™
B60SHTRG050090VPS |
5 to 9 |
Each for $37.57
Pack of 6 Each for $225.42
|
|
|||||
|
13640517
|
Fisherbrand™
B60SHTRG055080VPS |
5.5 to 8 |
Each for $37.57
Pack of 6 Each for $225.42
|
|
|||||
|
13640518
|
Fisherbrand™
B60SHTRG065130VPS |
6.5 to 13 |
Each for $33.94
Pack of 6 Each for $203.64
|
|
|||||
Description
Laboratory grade plastic pH strips in durable and easy to open flip-top vials
- Easy-to-use — simply dip strip in solution to be tested and match to color chart immediately
- Single color match with full 0.5 in. (1.27cm) matching area for quick and accurate matches
- Flip-top vial stays closed when dropped, but is easy to open with one-hand
- Sturdy vial keeps strips safe from the elements
- Full pH range (0.0-14.0) is available in a universal pH strip and finer measurements across the pH range
- 100 strips per vial; pack of 6 vials
Order Info
Box 6 Vials, 100 strips per vial.
Specifications
| For quick and accurate matches | |
| 100 Strips/Vial, 6 Vials/Pk. | |
| Room Temperature | |
| pH Strip |