missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Permaloc™ Formalin Containers
Description
- Patented double-clicking lock design
- 1st audible“click”ensures closure
- 2nd audible“click”achieves full 95kPa certification
- Available in two convenient sizes –20mL (10mL filled) neutral-buffered formalin;60mL (30mL filled) with wide mouth (53mm) opening neutral-buffered formalin
- Leak tested and independently 95kPa certified
- Avoid employee/patient exposure to hazardous chemicals
- Improve efficiency of specimen transport to the laboratory
- Economical bulk packaging with inner convenient transport packs help grow your Outreach Programs
Specifications
Specifications
| Product Type | Formalin Container |
| Capacity (Metric) | 40 mL, pre-filled 20 mL |
| Closure Type | Double clicking lock design |
| No. per Case | 480/Cs. |