Learn More
Fisherbrand™ Pasteur Pipets
Being packaged in strong corrugated material provides maximum protection from external shock for Fisherbrand™ Pasteur Pipets. Each pack also contains a foam pad to protect the pipet tips.
Description
Being packaged in strong corrugated material provides maximum protection from external shock for Fisherbrand™ Pasteur Pipets. Each pack also contains a foam pad to protect the pipet tips.
- Straight-cut tips and smooth, fire-polished tops to facilitate bulb attachment
- Positive constriction approximately 1 in. from the top to prevent cotton plugs from slipping into pipet barrel
- Easel configuration of each pack keeps pipets orderly and easily accessible
Specifications
| 0.27 in. | |
| Borosilicate Glass | |
| 1000 Case | |
| Smooth fire polished | |
| 22.86 cm |
| 7 mm | |
| 9 in. | |
| Straight | |
| 2 mL | |
| Easel |